Skip to Content
Merck
All Photos(2)

Key Documents

GF22757935

Cobalt

wire reel, 0.2m, diameter 0.1mm, hard, 99.99+%

Synonym(s):

Cobalt, CO005085

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$522.00

$522.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$522.00

About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

$522.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 227-579-35

resistivity

6.24 μΩ-cm, 20°C

L × diam.

0.2 m × 0.1 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth M Unice et al.
Chemico-biological interactions, 216, 53-74 (2014-04-15)
An updated biokinetic model for human exposures to cobalt (Co) was developed based on a comprehensive set of human pharmacokinetics data collected from five male and five female volunteers who ingested ∼1 mg Co/day of a Co supplement for 3
Dennis J Paustenbach et al.
Critical reviews in toxicology, 43(4), 316-362 (2013-05-10)
Cobalt (Co) is an essential element with ubiquitous dietary exposure and possible incremental exposure due to dietary supplements, occupation and medical devices. Adverse health effects, such as cardiomyopathy and vision or hearing impairment, were reported at peak blood Co concentrations
John J Devlin et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 9(4), 405-415 (2013-11-14)
Safety concerns regarding cobalt-containing metal alloy hip prosthetics (Co-HP) have resulted in product recalls, a medical device alert, and issuance of guidance for clinicians. Recently, cases of suspected prosthetic hip-associated cobalt toxicity (PHACT) from Co-HP have been reported. Although little
S Valanarasu et al.
Journal of nanoscience and nanotechnology, 14(6), 4286-4291 (2014-04-18)
Cobalt Oxide (Co3o4) thin films have been successfully coated onto glass substrates at various solution pH by sol-gel spin coating technique. The film thickness was estimated using weight gain method and it revealed that the film thickness increased with solution
K Praveena et al.
Journal of nanoscience and nanotechnology, 14(6), 4371-4376 (2014-04-18)
The Cobalt ferrite (CoFe2O4) powders were synthesized by Co-precipitation method. The as prepared ferrite powders were incorporated into a polyaniline matrix at various volumetric ratios. The as prepared composites of ferrite and polyaniline powders were characterized using X-ray diffraction (XRD)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service