GF21273937
Polymethylpentene
TPX®, sheet, thickness 12.7 mm, size 150 × 150 mm, natural
Synonym(s):
ME313415
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
sheet
manufacturer/tradename
Goodfellow 212-739-37
size
150 × 150 mm
thickness
12.7 mm
color
natural
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
TPX is a registered trademark of Mitsui Petrochemical Industries, Ltd.
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service