GF20999133
Hastelloy® C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
foil, 1m coil, 0.075mm thickness, hard
Synonym(s):
NI140271
Select a Size
$1,710.00
Select a Size
About This Item
$1,710.00
form
foil (coil - hard)
manufacturer/tradename
Goodfellow 209-991-33
size × thickness
1 m × 0.075 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service