GF19267634
Nickel-chromium alloy, Ni80Cr20
foil, 10m coil, 10mm coil width, 0.10mm thickness, as rolled
Synonym(s):
NI050236
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
temper (as rolled)
form
foil (coil)
manufacturer/tradename
Goodfellow 192-676-34
L × W × thickness
10 m × 10 mm × 0.10 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service