Skip to Content
Merck
All Photos(2)

Key Documents

GF17514312

Rhodium

foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%

Synonym(s):

Rhodium, RH000160

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,180.00

$1,180.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$1,180.00

About This Item

Empirical Formula (Hill Notation):
Rh
CAS Number:
Molecular Weight:
102.91
MDL number:
UNSPSC Code:
12141738
PubChem Substance ID:
NACRES:
NA.23

$1,180.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.90%

form

foil

manufacturer/tradename

Goodfellow 175-143-12

resistivity

4.33 μΩ-cm, 20°C

size × thickness

25 x 25 mm × 0.00025 mm

bp

3727 °C (lit.)

mp

1966 °C (lit.)

density

12.41 g/cm3 (lit.)

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimir V Grushin
Accounts of chemical research, 43(1), 160-171 (2009-10-01)
Although springing from two established fields, organometallic chemistry and fluorine chemistry, organometallic fluorine chemistry is still in its early stages. However, developments in this area are expected to provide new tools for the synthesis of selectively fluorinated organic compounds that
Pablo Etayo et al.
Chemical Society reviews, 42(2), 728-754 (2012-11-08)
During the last few decades, rhodium-catalysed asymmetric hydrogenation of diverse alkene classes has emerged as a powerful synthetic tool in the pharmaceutical industry, contributing to the manufacturing of chiral drugs, recent drug candidates for clinical trials, and major synthetic precursors
Feng Shi et al.
Topics in current chemistry, 292, 123-164 (2010-01-01)
The catalytic activation of a C-H bond is a fundamentally important organic transformation. There are now numerous reports of palladium-mediated C-H activation by the through-space interaction of a palladium center with a neighboring C-H bond. This type of C-H activation
Huw M L Davies et al.
Chemical Society reviews, 40(4), 1857-1869 (2011-03-02)
This tutorial review presents a description of the controlling elements of intermolecular C-H functionalization by means of C-H insertion by donor/acceptor rhodium carbenes. These rhodium carbenes, readily derived from the combination of diazo compounds with dirhodium(ii) catalysts, are sufficiently reactive
Jan Meine Ernsting et al.
Magnetic resonance in chemistry : MRC, 42(9), 721-736 (2004-08-13)
Rhodium is used for a number of large processes that rely on homogeneous rhodium-catalyzed reactions, for instance rhodium-catalyzed hydroformylation of alkenes, carbonylation of methanol to acetic acid and hydrodesulfurization of thiophene derivatives (in crude oil). Many laboratory applications in organometallic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service