Skip to Content
Merck
All Photos(2)

Key Documents

GF16662731

Tin

foil, 50mm disks, thickness 0.25mm, as rolled, 98.8%

Synonym(s):

Tin, SN000391

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

98.80%

form

foil

manufacturer/tradename

Goodfellow 166-627-31

resistivity

11 μΩ-cm, 20°C

diam. × thickness

50 mm × 0.25 mm

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF25742943GF34164075GF34214464
Tin foil, 50mm disks, thickness 0.25mm, as rolled, 98.8%

GF16662731

Tin

Tin foil, 100x100mm, thickness 0.25mm, as rolled, 98.8%

GF25742943

Tin

Tin foil, 10mm disks, thickness 0.25mm, as rolled, 98.8%

GF34164075

Tin

Tin foil, 4mm disks, thickness 0.25mm, as rolled, 98.8%

GF34214464

Tin

form

foil

form

foil

form

foil

form

foil

assay

98.80%

assay

98.80%

assay

98.80%

assay

98.80%

resistivity

11 μΩ-cm, 20°C

resistivity

11 μΩ-cm, 20°C

resistivity

11 μΩ-cm, 20°C

resistivity

11 μΩ-cm, 20°C

density

7.310 g/mL at 25 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

mp

231.9 °C (lit.)

mp

231.9 °C (lit.)

mp

231.9 °C (lit.)

mp

231.9 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mercury contamination removed with tin foil.
R Grajower et al.
Operative dentistry, 9(3), 101-104 (1984-01-01)
Gabriel Santpere et al.
Genome biology and evolution, 7(6), 1490-1505 (2015-05-16)
We set out to investigate potential differences and similarities between the selective forces acting upon the coding and noncoding regions of five different sets of genes defined according to functional and evolutionary criteria: 1) two reference gene sets presenting accelerated
M R Krigman et al.
Neurotoxicology, 5(2), 129-139 (1984-01-01)
The toxicology of tin is almost entirely the toxicology of the organic compounds of tin, for the metal itself and its inorganic compounds appear to be nearly harmless for practical purposes. Furthermore, the neurotoxicity of organotin is essentially that of
Are nickel, vanadium, silicon, fluorine, and tin essential for man? A review.
F H Nielsen et al.
The American journal of clinical nutrition, 27(5), 515-520 (1974-05-01)
Heinz Rüdel
Ecotoxicology and environmental safety, 56(1), 180-189 (2003-08-14)
This article reviews the literature related to the bioavailability of tin, inorganic tin compounds, and organotin compounds. On the one hand, the toxicity of metallic tin and inorganic tin compounds is low. In aqueous systems, the potential bioavailability of tin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service