GF15850267
Nickel-chromium alloy, Ni80Cr20
foil, 25 x 25mm, 0.18mm thickness, as rolled
Synonym(s):
NI050270
About This Item
form
foil (temper - as rolled)
manufacturer/tradename
Goodfellow 158-502-67
size × thickness
25 mm × 25 mm × 0.18 mm
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | GF01651115 | GF05035755 | GF24357629 |
---|---|---|---|
Goodfellow 158-502-67 | Goodfellow 016-511-15 | Goodfellow 050-357-55 | Goodfellow 243-576-29 |
25 mm × 25 mm × 0.18 mm | 100 mm × 100 mm × 0.18 mm | 100 mm × 100 mm × 0.125 mm | 50 mm × 50 mm × 0.25 mm |
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service