GF15485070
Tantalum
foil, 50mm disks, thickness 0.0125mm, 99.9%
Synonym(s):
Tantalum, TA000300, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.90%
form
foil
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 154-850-70
resistivity
13.5 μΩ-cm, 20°C
diam. × thickness
50 mm × 0.0125 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
PloS one, 8(6), e66447-e66447 (2013-06-27)
We report a simple method to fabricate nano-porous tantalum oxide films via anodization with Ta foils as the anode at room temperature. A mixture of ethylene glycol, phosphoric acid, NH4F and H2O was used as the electrolyte where the nano-porous
Observations on the use of tantalum foil in peripheral nerve surgery.
Journal of neurosurgery, 4(1), 69-71 (1947-01-01)
The use of tantalum foil as a mechanical block for the separation of spinal cord and brain in chick embryos.
The Anatomical record, 94, 355-355 (1946-03-01)
Journal of the American Chemical Society, 131(34), 12048-12049 (2009-08-27)
A rutile microstructure with a novel cross-medal morphology was prepared via a hydrothermal method by using a Tantalum foil as both the doping source and the substrate. This rutile cross-medal prefers to grow along an unprecedented preferential orientation of <110>
Use of a tantalum foil tube to maintain patency of the nasofrontal duct.
A.M.A. archives of otolaryngology, 54(2), 167-171 (1951-08-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service