Skip to Content
Merck
All Photos(2)

Key Documents

GF08081670

Lead

wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%

Synonym(s):

Lead, PB005130

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$651.00

$651.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$651.00

About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

$651.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 080-816-70

resistivity

20.65 μΩ-cm

L × diam.

2 m × 1.0 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paraskevi Malea et al.
Ecotoxicology and environmental safety, 104, 175-181 (2014-04-01)
The effects of lead uptake on microtubule integrity and cell viability in intermediate-juvenile leaf blades of the seagrass Cymodocea nodosa were investigated under laboratory conditions in increasing exposure concentrations (0.1, 0.25, 0.5, 5, 10, 20 and 40mg/L). Uptake kinetics was
Yunbiao Wang et al.
Ecotoxicology and environmental safety, 104, 317-322 (2014-04-15)
In the nematode Caenorhabditis elegans, stress resistance can be regulated by dauer formation (daf) genes. In the present study, regulation of heavy metal lead (Pb) toxicity by the 90-kDa heat shock proteins (Hsp90; daf-21) was investigated in both wild-type C.
Guillermo Horta-Puga et al.
Bulletin of environmental contamination and toxicology, 92(5), 567-573 (2014-02-11)
Lead (Pb) pollution history (1855-2001 A.D.) of the southern Gulf of Mexico (SGM) was reconstructed from the geochemical record contained in the annual bands of the hermatypic coral Orbicella faveolata from the Veracruz Reef System, Mexico. Pb concentrations ranged from
Jun Yoshinaga et al.
Environmental pollution (Barking, Essex : 1987), 189, 223-228 (2014-04-01)
The levels of 25 elements in house dust collected from 100 general Japanese residences were measured. Factor analysis was applied on the multi-element data to explore source of Pb (median concentration 49.1 mg/kg) in house dust. Six factors were extracted
Yi Yang et al.
Toxicology letters, 227(3), 172-178 (2014-04-09)
Lead is a toxic heavy metal with many recognized adverse health side effects. The central nervous system is the main target of lead toxicity. Although many studies on lead toxicity were conducted, the mechanism of lead toxicity remains uncertain. One

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service