Skip to Content
Merck
All Photos(2)

Key Documents

GF08067214

Iron

foil, 100x100mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%

Synonym(s):

Iron, FE000465

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

Assay

99.8%

form

foil

manufacturer/tradename

Goodfellow 080-672-14

resistivity

9.71 μΩ-cm

L × W × thickness

100 mm × 100 mm × 2.0 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Won Jang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 59(12), 2503-2507 (2009-06-23)
Zero valent iron has been successfully used for the degradation of a wide range of contaminants. However, this reaction of using ZVI particle produces a large quantity of iron sludge. To solve the problem, we report the synthesis of self-organized
Qiu et al.
Physical review letters, 85(7), 1492-1495 (2000-09-06)
FeF (2) films are grown by the reaction of XeF (2) and SeF (6) with iron foil. The growth initially follows the Mott-Cabrera parabolic rate law, indicating that the process is diffusion limited. At a certain film thickness, however, the
Arnold L Demain et al.
Applied microbiology and biotechnology, 73(1), 55-59 (2006-04-20)
When tetanus toxin is made by fermentation with Clostridium tetani, the traditional source of iron is an insoluble preparation called reduced iron powder. This material removes oxygen from the system by forming FeO(2) (rust). When inoculated in a newly developed
Florence Lima et al.
Environmental and molecular mutagenesis, 55(3), 266-277 (2014-04-12)
DNA methylation is an epigenetic mechanism that drives phenotype and that can be altered by environmental exposures including radiation. The majority of human radiation exposures occur in a relatively low dose range; however, the biological response to low dose radiation
Antonino Missineo et al.
Infection and immunity, 82(6), 2448-2459 (2014-04-02)
Staphylococcus lugdunensis is a coagulase-negative staphylococcus that is a commensal of humans and an opportunistic pathogen. It can cause a spectrum of infections, including those that are associated with the ability to form biofilm, such as occurs with endocarditis or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service