Skip to Content
Merck
All Photos(2)

Key Documents

GF03146318

Nickel

rod, 1.5 mm diameter, length 500 mm, purity 99%

Synonym(s):

NI007910

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$299.00

$299.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$299.00

About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

$299.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 031-463-18

resistivity

6.97 μΩ-cm, 20°C

L

500 mm

diameter

1.5 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service