Skip to Content
Merck
All Photos(2)

Key Documents

GF01141197

Molybdenum

rod, 200mm, diameter 3.0mm, centerless ground, 99.9%

Synonym(s):

Molybdenum, MO007901

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.9%

form

rod

manufacturer/tradename

Goodfellow 011-411-97

resistivity

5.0 μΩ-cm, 20°C

L × diam.

200 mm × 3.0 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad-Reza Rashidi et al.
Expert opinion on drug metabolism & toxicology, 6(2), 133-152 (2010-01-26)
Molybdenum hydroxylases, aldehyde oxidase and xanthine oxidase, are metalloflavoproteins that catalyze both oxidation and reduction of a broad range of drugs and other xenobiotics indicating the importance of these enzymes in drug oxidation, detoxification and activation. Both enzymes are also
Maria Lyra et al.
Hellenic journal of nuclear medicine, 14(1), 49-55 (2011-04-23)
The sleeping giant of molybdenum-99 ((99)Mo) production is grinding to a halt and the world is wondering how this happened. Fewer than 10 reactors in the world are capable of producing radio nuclides for medicine; approximately 50% of the world's
Ralf R Mendel
The Journal of biological chemistry, 288(19), 13165-13172 (2013-03-30)
The transition element molybdenum needs to be complexed by a special cofactor to gain catalytic activity. Molybdenum is bound to a unique pterin, thus forming the molybdenum cofactor (Moco), which, in different variants, is the active compound at the catalytic
Viswanathan S Saji et al.
ChemSusChem, 5(7), 1146-1161 (2012-06-14)
The electrochemical behaviors of molybdenum and its oxides, both in bulk and thin film dimensions, are critical because of their widespread applications in steels, electrocatalysts, electrochromic materials, batteries, sensors, and solar cells. An important area of current interest is electrodeposited
Li Lv et al.
Journal of nanoscience and nanotechnology, 14(5), 3521-3526 (2014-04-17)
In this work, we report the size and shape-controlled synthesis of CaMoO4 nanocrystals with an aim of investigate their structural, electronic, and luminescent properties. The samples were carefully characterized by X-ray diffraction, transmission electron microscopy, Fourier transformed infrared spectroscopy, UV-vis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service