Skip to Content
Merck
All Photos(2)

Key Documents

GF00761217

Titanium

wire reel, 200m, diameter 0.25mm, as drawn, 99.6+%

Synonym(s):

Titanium, TI005123

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,270.00

$1,270.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$1,270.00

About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

$1,270.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

≥99.6%

form

wire

autoignition temp.

860 °F

manufacturer/tradename

Goodfellow 007-612-17

resistivity

42.0 μΩ-cm, 20°C

L × diam.

200 m × 0.25 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaoming Yang et al.
Journal of nanoscience and nanotechnology, 14(6), 4292-4296 (2014-04-18)
The structural and functional characterizations of titanium nitride (TiN) advanced ceramic microsprings (CMSs), with a coil diameter of several micrometers and synthesized by chemical vapor deposition (CVD) were investigated by microscopy techniques. The CMSs were sufficiently mechanically elastic for extension
Konstantinos X Michalakis et al.
The Journal of oral implantology, 40(2), 146-152 (2014-05-02)
Dental implants with an internal connection have been designed to establish a better stress distribution when lateral external forces act on the prosthesis and minimize the forces transmitted to the fastening screw. In the present study, 10 externally and 10
P F Du et al.
Journal of nanoscience and nanotechnology, 14(6), 4164-4169 (2014-04-18)
Novel TiO2 nanoparticles/nanofibers (NPs/NFs) bilayered nano-composite photoanode film for dye-sensitized solar cells (DSSCs) was fabricated through the combination of spin-coating and electrospinning. The NPs and NFs layers have complementary roles. The underlaid spin-coated NPs layer provides the photoanode film with
Cédric Weber et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(16), 5790-5795 (2014-04-11)
We carry out a first-principles atomistic study of the electronic mechanisms of ligand binding and discrimination in the myoglobin protein. Electronic correlation effects are taken into account using one of the most advanced methods currently available, namely a linear-scaling density
Nikiwe Mhlanga et al.
Journal of nanoscience and nanotechnology, 14(6), 4269-4277 (2014-04-18)
This study reports the synthesis, characterisation and thermal properties of polylactide (PLA)/titanium dioxide nanoparticles (TiO2 NPs) composites using the sol-gel method. The percentage weight of TiO2 NP sol was varied from 3, 8, 11 and 14. The synthesised composites were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service