Skip to Content
Merck
All Photos(1)

Key Documents

G2834

Sigma-Aldrich

L-Glutamic acid monosodium salt monohydrate

98%

Synonym(s):

L-2-Aminopentanedioic acid, MSG, Sodium L-glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
187.13
Beilstein:
4164348
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D +21.6°, c = 1 in 6 M HCl

mp

232 °C (dec.) (lit.)

SMILES string

O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O

InChI

1S/C5H9NO4.Na.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1

InChI key

GJBHGUUFMNITCI-QTNFYWBSSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service