Skip to Content
Merck
All Photos(1)

Key Documents

F15145

Sigma-Aldrich

Bis(4-fluorophenyl) sulfone

99%

Synonym(s):

4,4′-Difluorodiphenyl sulfone, 4-Fluorophenyl sulfone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$239.00

$239.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$239.00

About This Item

Linear Formula:
(FC6H4)2SO2
CAS Number:
Molecular Weight:
254.25
Beilstein:
1970911
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$239.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

98-100 °C (lit.)

SMILES string

Fc1ccc(cc1)S(=O)(=O)c2ccc(F)cc2

InChI

1S/C12H8F2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H

InChI key

PLVUIVUKKJTSDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Fan Zhao et al.
Journal of colloid and interface science, 448, 380-388 (2015-03-11)
Here we describe the development of versatile antifouling polyethersulfone (PES) filtration membranes modified via surface grafting of zwitterionic polymers from a reactive amphiphilic copolymer additive. Amphiphilic polyethersulfone-block-poly(2-hydroxyethyl methacrylate) (PES-b-PHEMA) was beforehand designed and used as the blending additive of PES

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service