Skip to Content
Merck
All Photos(1)

Key Documents

F1204

Sigma-Aldrich

Fluorene-1-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$69.90

$69.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$69.90

About This Item

Empirical Formula (Hill Notation):
C14H10O2
CAS Number:
Molecular Weight:
210.23
Beilstein:
2330772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$69.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

246-248 °C (lit.)

SMILES string

OC(=O)c1cccc-2c1Cc3ccccc-23

InChI

1S/C14H10O2/c15-14(16)12-7-3-6-11-10-5-2-1-4-9(10)8-13(11)12/h1-7H,8H2,(H,15,16)

InChI key

HTPXFGUCAUTOEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Kemnitzer et al.
Bioorganic & medicinal chemistry letters, 20(3), 1288-1292 (2009-12-26)
As a continuation of our studies of apoptosis inducing 9-oxo-9H-fluorene-1-carboxamides as potential anticancer agents, we explored modification of the 9-oxo-9H-fluorene ring. SAR studies showed that most changes to the 9-oxo-9H-fluorene ring were not well tolerated, except the 9H-fluorene (2b) and
A C Blackburn et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 4), 907-910 (1996-04-15)
In fluorene-1-carboxylic acid, C14H10O2, the sole hydrogen bond is of the cyclic dimer type about a center of symmetry. The carboxyl H atom is ordered. Distances in the fluorene core are very similar to those in fluorene itself; the fluorene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service