Skip to Content
Merck
All Photos(1)

Key Documents

E49207

Sigma-Aldrich

2-Ethylthiophene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$272.00

$272.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$272.00

About This Item

Empirical Formula (Hill Notation):
C6H8S
CAS Number:
Molecular Weight:
112.19
Beilstein:
105402
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$272.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.513 (lit.)

bp

132-134 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CCc1cccs1

InChI

1S/C6H8S/c1-2-6-4-3-5-7-6/h3-5H,2H2,1H3

InChI key

JCCCMAAJYSNBPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

85.3 °F - closed cup

Flash Point(C)

29.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service