DWKW224585
WHEATON® clear sample vial with PTFE faced rubber lined cap packed in partitioned tray
glass, tube capacity (12 mL), screw cap
Synonym(s):
Clear Sample Vial, PTFE-Lined Vial, Sample Vial, W224585
Select a Size
$373.00
Select a Size
About This Item
$373.00
Recommended Products
material
flat bottom
glass
solid black phenolic cap
transparent borosilicate glass tube
sterility
non-sterile
feature
with cap
packaging
case of 144 ea partitioned tray
manufacturer/tradename
WHEATON® W224585
tube capacity
(12 mL)
tube diam.
(19 mm)
fitting
neck (External Thread)
General description
- Screw caps are attached to vials.
- It is suggested to use rubber lined caps for aqueous samples, and PTFE faced rubber lined caps for organic samples, and metal foil liner for hydrocarbons.
- Vials and caps are autoclavable.
- 2 mL vials fit the M-T Vial File, 228778.
- 4 mL vials fit the M-T Vial File, 228780.
- Vials with caps attached packaged in partitioned trays
Features and Benefits
- Clear vials are converted from Type I borosilicate glass tubing
- Amber vials are converted from amber borosilicate glass tubing that conforms to USP Type I requirements for light transmission to protect light sensitive products
- Sample vials with caps attached to vial help maintain cleanliness
- Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
- Shorty Vial® feature a low profile for greater stability and less dead volume
- 14B rubber lined black phenolic screw cap for aqueous samples
- PTFE / 14B rubber lined black phenolic screw cap for organic samples and improved chemical compatibility
- Replacement caps can be purchased separately
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)