DWKCTA0050P
Azlon Hexagonal Base PMP Cylinders with Printed Graduations
class B, capacity 50 mL , with +/-0.5mL tolerance
Synonym(s):
CTA0050P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121805
NACRES:
NA.54
feature
class B
graduated
packaging
pkg of 2 ea
manufacturer/tradename
Azlon CTA0050P
capacity
50 mL , with +/-0.5mL tolerance
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for the measurement and dispensing of liquids in the laboratory
Features and Benefits
Calibrated to ISO 6706 and DIN 12681 class A tolerances
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service