Skip to Content
Merck
All Photos(1)

Key Documents

DWKBGE242P

Azlon Square Bottles

graduated, cap, Plug, translucent high-density polyethylene, capacity 2000 mL

Synonym(s):

BGE242P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43
Pricing and availability is not currently available.

material

translucent high-density polyethylene

feature

cap Plug
graduated

packaging

pkg of 5 ea

manufacturer/tradename

Azlon

capacity

2000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for storage of liquid and powder samples

Features and Benefits

Space saving square design

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service