Key Documents
DWK92812F-250
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper
glass flask, flask capacity (250 mL), class A
Select a Size
6 EA
$621.00
$621.00
Estimated to ship onApril 14, 2025
Select a Size
Change View
6 EA
$621.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$621.00
Estimated to ship onApril 14, 2025
material
PTFE stopper
glass flask
feature
class A
packaging
case of 6 ea
manufacturer/tradename
Kimble® 92812F-250
flask capacity
(250 mL)
General description
Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Features and Benefits
• Manufactured from borosilicate glass 3.3• Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets• Heavy duty walls provide superior strength, durability and safety• Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills• Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets• Printed large volume numbers allow quick identification when selecting glassware• Flat interior bottoms for stirring with standard stir bars• Calibrate to contain• Graduated to Class A volumetric tolerances•
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)