Key Documents
DWK92812F-20
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper
glass flask, flask capacity (20 mL), class A
Select a Size
6 EA
$388.00
$388.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
6 EA
$388.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$388.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PTFE stopper
glass flask
feature
class A
packaging
case of 6 ea
manufacturer/tradename
Kimble® 92812F-20
flask capacity
(20 mL)
General description
Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Features and Benefits
  • Manufactured from borosilicate glass 3.3
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Heavy duty walls provide superior strength, durability and safety
  • Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Printed large volume numbers allow quick identification when selecting glassware
  • Flat interior bottoms for stirring with standard stir bars
  • Calibrate to contain
  • Graduated to Class A volumetric tolerances
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Can a 1-mL pipette fit into the neck of this volumetric flask?
    1 answer
    1. Yes a 1-mL pipette can fit into the neck of this volumetric flask.
      Helpful?