Key Documents
DWK73808-28400
Kimble® Closed Top Polypropylene Screw Thread Cap With Welded Ptfe-Faced Silicone Liner
28-400
Select a Size
1 PKG
$132.00
$132.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 PKG
$132.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
$132.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PTFE/silicone liner
red polypropylene screw cap (Push fit)
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 73808-28400
fitting
thread
Application
Closure solution for vials and bottles
Features and Benefits
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Closures are ideal for repeated autoclaving
  • Welded liner technology eliminates the possibility of glue contamination
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)