Key Documents
DWK73785-50
KIMBLE® centrifuge tube
with screw-top finish, conical bottom clear glass, capacity (50 mL), without cap, non-sterile
Select a Size
72 EA
$215.00
$215.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
72 EA
$215.00
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
$215.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
conical bottom clear glass
conical bottom
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
packaging
case of 72 ea Modular trays are shrink-wrapped
manufacturer/tradename
Kimble® 73785-50
capacity
(50 mL)
fitting
neck (External Thread)
General description
Screw Thread Disposable Centrifuge Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)