Key Documents
DWK73665-16
KIMBLE® KIM-KAP® push cap
blue polypropylene cap, cap diam. (16 mm)
Select a Size
1000 EA
$187.00
$187.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1000 EA
$187.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$187.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
blue polypropylene cap
sterility
non-sterile
packaging
case of 1000 ea
manufacturer/tradename
Kimble® 73665-16
cap diam.
(16 mm)
General description
  • KIM-KAP polypropylene closures for culture tubes
  • Autoclavable
  • Color coded
Features and Benefits
  • Autoclavable
  • Color coded
Legal Information
KIM-KAP is a registered trademark of DWK LIFE SCIENCES LLC
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)