Key Documents
DWK62113D-312
Kimble® 33 Expansion Tubular Borosilicate Glass Serum Vial Without Closure
Select a Size
1 PKG
$1,330.00
$1,330.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 PKG
$1,330.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.43
$1,330.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
Borosilicate glass/PTFE tube
tubulated bottom
feature
closure type: no
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 62113D-312
General description
Autoclavable serum vials are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, culture media, chromatography samples, and more.
Application
Storage and transport of samples, lyophilization, contain harsh reagents or liquids.
Features and Benefits
  • Special design provides extra strength for freeze-drying applications
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Aluminum seals, stoppers and septa are available separately
  • Converted from Type I borosilicate glass tubing, vials conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)