Skip to Content
Merck
All Photos(6)

Key Documents

DWK292807309

Duran® Gl 45 Bottle Carrying System

Synonym(s):

292807309

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EACH
$204.00

About This Item

UNSPSC Code:
41120000
NACRES:
NB.17

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

yellow polypropylene

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 292807309

compatibility

use with DURAN® Original GL 45 Bottle

Looking for similar products? Visit Product Comparison Guide

General description

The Bottle Carrying System range fits standard laboratory bottles that have a GL 45 neck finish, and are available for four bottle sizes: 2, 5, 10 or 20 litres. The carriers are ergonomically designed, and offer health and safety benefits to the user. They allow the easy and safe transport of bottles around the laboratory or production facility. The robust pouring side handle on the 5 L version makes pouring liquids from the larger bottles more controllable and safer. All materials are fully autoclavable. The collar that fits to the neck of the bottle is only removable by cutting, so the attachment is 100 % secure in use.

Application

The new modular Bottle Carrying System fi ts standard laboratorybottles that have a GL 45 neck fi nish and is compatible with fourbottle sizes 2, 5, 10 and 20 litres. It is ergonomically designed, ando ers health & safety benefi ts to the user and allows easy and safetransport of bottles around the laboratory or production facility.

Features and Benefits

All materials are fully autoclavable.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service