DWK241935702
DURAN® Erlenmeyer flask with standard ground joint
flask capacity 1,000 mL, neck joint: ST/NS 45/40
Synonym(s):
241935702
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3 flask
flat
Joint
joint: ST/NS neck45/40
Sterilization Compatibility
autoclavable compatible
feature
without cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 241935702
flask capacity
1,000 mL
General description
Tight closure due to standard ground joint. Fired-on, highly durable white print. Easy mixing due to conical form
Other Notes
Browse all Erlenmeyer flasks
Legal Information
DURAN is a registered trademark of DWK Life Sciences