Skip to Content
Merck
All Photos(1)

Key Documents

DWK218018855

DURAN® graduated laboratory bottle

with cap, round glass bottle, bottle capacity (15,000 mL), non-sterile

Synonym(s):

218018855

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$799.00

$799.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EACH
$799.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

$799.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

round glass bottle

sterility

non-sterile

feature

with cap

packaging

case of 1 ea

manufacturer/tradename

DURAN® 218018855

bottle capacity

(15,000 mL)

Looking for similar products? Visit Product Comparison Guide

General description

with DIN 168-1 Thread, graduated

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service