Skip to Content
Merck
All Photos(2)

Key Documents

DWK213543804

DURAN® powder funnel with short wide stem

glass funnel(s), funnel capacity (50 mL)

Synonym(s):

213543804

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
$268.00

$268.00


Estimated to ship onMay 24, 2025



Select a Size

Change View
10 EACH
$268.00

About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

$268.00


Estimated to ship onMay 24, 2025


material

glass funnel(s)

packaging

case of 10 ea

manufacturer/tradename

DURAN® 213543804

funnel capacity

(50 mL)

Looking for similar products? Visit Product Comparison Guide

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service