Skip to Content
Merck
All Photos(1)

Key Documents

DWK211947353

Duran® Filtering Flasks

Bottle shape, WITH KECK ASSEMBLY SET

Synonym(s):

211947353

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$373.00

$373.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EACH
$373.00

About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

$373.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 211947353

capacity

5000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Separations by vacuum filtration, Filtration at reduced pressure, Filtrieren, Vacuum filtration, Safe working under vacuum, Boro 3.3, PBT, PP, VMQ, EPDM

Features and Benefits

Vacuum resistant due to the heavy wall thickness; These filtering flasks fulfil the regulations of the equipment and product safety regulations; Fired-on, highly durable white print; Fired-on, highly durable white print;

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is there a matching rubber stopper/bung to fit this flask? preferably with a 15mm centre hole?

    1 answer
    1. This vessel will accommodate a #13.5 stopper, but this item is not available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service