Key Documents
DWK11-0056K
Wheaton® Microliter Snap Caps With Septa
black PTFE/silicone cap
Select a Size
100 EACH
$86.80
$86.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
100 EACH
$86.80
About This Item
UNSPSC Code:
41100000
NACRES:
NB.11
$86.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
black PTFE/silicone cap
feature
cap pre-cut
cap starburst
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® 11-0056K
Application
Chromatography
Features and Benefits
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)