Skip to Content
Merck
All Photos(1)

Key Documents

D99250

Sigma-Aldrich

Diethyl 2,2-diethoxyethylphosphonate

95%

Synonym(s):

Diethyl phosphonoacetaldehyde diethyl acetal, NSC 407851, NSC 77101

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$304.00

$304.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$304.00

About This Item

Linear Formula:
(CH3CH2O)2CHCH2P(O)(OCH2CH3)2
CAS Number:
Molecular Weight:
254.26
Beilstein:
1785614
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$304.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.430 (lit.)

bp

146-149 °C/14 mmHg (lit.)

density

1.052 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(CP(=O)(OCC)OCC)OCC

InChI

1S/C10H23O5P/c1-5-12-10(13-6-2)9-16(11,14-7-3)15-8-4/h10H,5-9H2,1-4H3

InChI key

LUQYELQXRPNKRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • α,β-Alkenal derivatives by two-carbon homologation
  • Lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions
  • α-Phosphovinyl radicals via a radical trapping sequence
  • Inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions

Reactant for Friedel-Crafts reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service