Skip to Content
Merck
All Photos(1)

Documents

D91071

Sigma-Aldrich

Diethyl benzylphosphonate

99%

Synonym(s):

(Diethoxyphosphonomethyl)benzene, Benzylphosphonic acid diethyl ester, NSC 62294

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
228.22
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.497 (lit.)

bp

106-108 °C/1 mmHg (lit.)

density

1.095 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(Cc1ccccc1)OCC

InChI

1S/C11H17O3P/c1-3-13-15(12,14-4-2)10-11-8-6-5-7-9-11/h5-9H,3-4,10H2,1-2H3

InChI key

AIPRAPZUGUTQKX-UHFFFAOYSA-N

Related Categories

Application

Reactant for synthesis of:
  • 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders
  • Natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions
  • Stilbenes via on-column oxidation of vicinal diols and Horner-Emmons reactions
  • Inhibitors of teh Wnt pathway for colon cancer repression using Wadsworth-Emmons reactions
  • Antimalarial drug analogs against P. falciparum

Reactant for:
  • Cyclization of aryl ethers, amines, and amides
  • Investigating the effects of functional groups on the performance of clue organic LEDs

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ananya Thomas et al.
Polymers, 12(8) (2020-08-17)
As a part of our ongoing investigations on passively fire protecting polymeric materials, we have been employing both reactive and additive routes involving phosphorus-containing compounds. These included inorganic and organic substances, and in the latter case, the phosphorus-bearing groups differed
Jack Saltiel et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 18(9), 2174-2179 (2019-05-28)
cis,trans-1,2-Dideuterio-1,4-diphenyl-1,3-butadiene (ct-DPBd2) was synthesized and its cis-trans photoisomerization in cyclohexane-d12 (C6D12) at room temperature was monitored by 1H NMR spectroscopy. The results reveal formation of only trans,trans-1,2-dideuterio-1,4-diphenyl-1,3-butadiene (tt-DPBd2). The failure to detect formation of trans,cis-1,2-dideuterio-1,4-diphenyl-1,3-butadiene (tc-DPBd2) eliminates the possibility that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service