Skip to Content
Merck
All Photos(1)

Key Documents

D8765

Sigma-Aldrich

Diethylcarbamazine citrate salt

≥98%

Synonym(s):

N,N-Diethyl-4-methyl-1-piperazinecarboxamide citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H21N3O · C6H8O7
CAS Number:
Molecular Weight:
391.42
Beilstein:
3836352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥98%

form

powder

antibiotic activity spectrum

parasites

Mode of action

enzyme | inhibits

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)C(=O)N1CCN(C)CC1

InChI

1S/C10H21N3O.C6H8O7/c1-4-12(5-2)10(14)13-8-6-11(3)7-9-13;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-9H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

PGNKBEARDDELNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koneni V Sashidhara et al.
Bioorganic & medicinal chemistry letters, 22(4), 1527-1532 (2012-01-31)
A series of 3,6-epoxy [1,5]dioxocines were synthesized and evaluated for their antifilarial activity against adult parasites of human lymphatic filarial parasite Brugia malayi (sub-periodic strain) in vitro. Out of these, six compounds (4a-f) possessed improved in vitro anti-filarial activity and
Ana Wládia Lima et al.
Revista da Sociedade Brasileira de Medicina Tropical, 45(6), 745-750 (2013-01-09)
The Global Programme to Eliminate Lymphatic Filariasis was launched with the goal of eliminating this disease via the annual mass drug administration (MDA) of a single dose of antifilarial drugs. Adverse drug reactions following MDA are a major factor of
Cathy Steel et al.
PLoS neglected tropical diseases, 6(12), e1940-e1940 (2012-12-14)
Antibody (Ab) to the Wuchereria bancrofti (Wb) infective larval (L3) antigen Wb123, using a Luciferase Immunoprecipitation System (LIPS) assay, has been shown to be a species-specific, early marker of infection developed for potential use as a surveillance tool following transmission
B K Singh et al.
European journal of medicinal chemistry, 43(12), 2717-2723 (2008-03-15)
A series of 2-sulfanyl-6-methyl-1,4-dihydropyrimidines (8-21) were synthesized in good yields by alkylation of 5-methyl-6-phenyl-2-thioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid ethyl esters (2-7) with different alkyl or aralkyl halides in the presence of a combination of anhydrous K(2)CO(3) and catalytic amount of tetrabutyl ammonium bromide.
Rabindra Nath Roy et al.
Journal of health, population, and nutrition, 31(2), 171-177 (2013-08-13)
India adopted WHO's strategy of repeated rounds of mass drug administration (MDA) with diethylcarbamazine to eliminate lymphatic filariasis. The present study attempted to assess the coverage and awareness of and compliance with MDA for elimination of lymphatic filariasis in Burdwan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service