Skip to Content
Merck
All Photos(1)

Key Documents

D7786

Sigma-Aldrich

Decylplastoquinone

~75%

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$194.00

$194.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$194.00

About This Item

Empirical Formula (Hill Notation):
C18H28O2
CAS Number:
Molecular Weight:
276.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$194.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Assay

~75%

form

liquid

storage temp.

−20°C

SMILES string

CCCCCCCCCCC1=CC(=O)C(C)=C(C)C1=O

InChI

1S/C18H28O2/c1-4-5-6-7-8-9-10-11-12-16-13-17(19)14(2)15(3)18(16)20/h13H,4-12H2,1-3H3

InChI key

APNQQQRDLMWNLM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam Paredes et al.
PloS one, 10(9), e0137472-e0137472 (2015-09-12)
The present work studies the effects of cold on photosynthesis, as well as the involvement in the chilling stress of chlororespiratory enzymes and ferredoxin-mediated cyclic electron flow, in illuminated plants of Hibiscus rosa-sinensis. Plants were sensitive to cold stress, as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service