Skip to Content
Merck
All Photos(2)

Key Documents

D7631

Sigma-Aldrich

5,5-Dimethyl-2,4-oxazolidinedione

Synonym(s):

DMO, Dimethadione, NSC 30152

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$229.00

$229.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$229.00

About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$229.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

form

powder

Quality Level

mp

77-80 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)OC(=O)NC1=O

InChI

1S/C5H7NO3/c1-5(2)3(7)6-4(8)9-5/h1-2H3,(H,6,7,8)

InChI key

JYJFNDQBESEHJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Synthesis of nickel benzoannulated or naphthoannulated β-oxatetraazachlorin complex
  • Being used as a cathepsin K inhibitor

Molecule involved in pharmacological studies including:
  • Identification of bioequivalent generic substitute for antiepileptic drugs
  • Screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysis
  • Dissolving protein plugs in patients with choledochal cyst / pancreaticobiliary maljunctions

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Danielsson et al.
Birth defects research. Part A, Clinical and molecular teratology, 79(8), 595-603 (2007-06-23)
The antiepileptic drugs (AEDs) phenytoin, phenobarbital, dimethadione, and carbamazepine cause a similar pattern of malformations in humans, with an increased risk after polytherapy. The teratogenicity has been linked to cardiac rhythm disturbances and hypoxic damage as a consequence of their
D P Nolan et al.
European journal of biochemistry, 267(15), 4601-4614 (2000-07-21)
Conditions for the use of both [14C]methylamine and 5, 5-dimethyl[14C]oxa-azolidine-2,4-dione (DMO) to measure the H+ concentration of intracellular compartments of monomorphic long thin bloodstream forms of Trypanosoma brucei were established. Neither probe was actively transported or bound to internal components
Cunxi Wang et al.
Regulatory toxicology and pharmacology : RTP, 81, 171-182 (2016-10-30)
Dicamba tolerant (DT) soybean, cotton and maize were developed through constitutive expression of dicamba mono-oxygenase (DMO) in chloroplasts. DMO expressed in three DT crops exhibit 91.6-97.1% amino acid sequence identity to wild type DMO. All DMO forms maintain the characteristics
Jingxuan Wang et al.
Food & function, 10(8), 4823-4833 (2019-07-19)
Donkey milk is considered to be a valuable nutritional source. Deeper knowledge of the constituents of donkey milk is necessary. As multifunctional components of milk, oligosaccharides have been reported to have the potential to support intestine development. We studied the
Hemal Mehta et al.
The British journal of ophthalmology, 102(4), 479-482 (2017-08-06)
To determine whether early vision gains predict long-term visual outcomes in the BEVORDEX randomised clinical trial of bevacizumab or dexamethasone implants for diabetic macular oedema. Post hoc analysis of 68 study eyes (77%) that completed 2 years follow-up of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service