Skip to Content
Merck
All Photos(2)

Key Documents

D71607

Sigma-Aldrich

4,5-Dichloro-o-phenylenediamine

97%

Synonym(s):

1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichloro-1,2-phenylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$155.00
25 G
$516.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$155.00
25 G
$516.00

About This Item

Linear Formula:
Cl2C6H2(NH2)2
CAS Number:
Molecular Weight:
177.03
Beilstein:
1072811
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

158-164 °C (lit.)

SMILES string

Nc1cc(Cl)c(Cl)cc1N

InChI

1S/C6H6Cl2N2/c7-3-1-5(9)6(10)2-4(3)8/h1-2H,9-10H2

InChI key

IWFHBRFJOHTIPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Zöllner et al.
Journal of mass spectrometry : JMS, 38(7), 709-714 (2003-08-05)
The derivatization reaction of the mycotoxin moniliformin with 1,2-diamino-4,5-dichlorobenzene was previously introduced to improve distinctly the sensitivity of an assay applying high-performance liquid chromatography prior to fluorescence detection. In the course of the implementation of this derivatization approach into a
Jin-Aa Oh et al.
Journal of separation science, 40(20), 3958-3968 (2017-08-24)
Malondialdehyde has been used as a biomarker for lipid peroxidation in biological samples. An ultra-high performance liquid chromatography with tandem mass spectrometry method was developed to determine the levels of malondialdehyde in human urine and saliva samples. To select the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service