Skip to Content
Merck
All Photos(1)

Key Documents

D68401

Sigma-Aldrich

2,4-Dichloro-1-nitrobenzene

97%

Synonym(s):

1,3-Dichloro-4-nitrobenzene, asym.-Nitro-m-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein:
1451655
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

bp

258 °C (lit.)

mp

29-32 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Cl)cc1Cl

InChI

1S/C6H3Cl2NO2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H

InChI key

QUIMTLZDMCNYGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Breinholt et al.
Cancer letters, 154(2), 201-210 (2000-05-12)
The administration of lycopene to female rats at doses ranging from 0.001 to 0.1 g/kg b.w. per day for 2 weeks was found to alter the drug-metabolizing capacity and antioxidant status of the exposed animals. An investigation of four cytochrome
Bruno Miguel Neves et al.
Toxicology letters, 177(1), 74-82 (2008-02-19)
The development of non-animal methods for skin sensitization testing is an urgent challenge. Some of the most promising in vitro approaches are based on the analysis of phenotypical and functional modifications induced by sensitizers in dendritic cell models. In this
Maja Aleksic et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(4), 723-733 (2007-02-24)
Covalent modification of skin proteins by electrophiles is a key event in the induction of skin sensitisation but not skin irritation although the exact nature of the binding mechanisms has not been determined empirically for the vast majority of sensitisers.
R M Bolton et al.
Analytical biochemistry, 216(2), 418-423 (1994-02-01)
Ammonium 4-chloro-7-sulfobenzofurazan (Sbf-Cl) is a water-soluble fluorescent reagent which is highly specific for thiol groups. We describe here a simple and sensitive fluorescence assay which is highly specific for subunit 3 of the rat glutathione S-transferases. Specific activities of isoenzymes
Teresa J Matos et al.
Journal of dermatological science, 39(2), 113-123 (2005-05-18)
The intracellular mechanisms involved in the activation of DCs during sensitization in allergic contact dermatitis (ACD) are not known. Here, we investigated the effect of a strong sensitizer, 2,4-dinitrofluorobenzene (DNFB) on the activity of MAPKs in a dendritic cell (DC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service