Skip to Content
Merck
All Photos(2)

Documents

D67200

Sigma-Aldrich

2,3-Dichloro-1,4-naphthoquinone

98%

Synonym(s):

Dichlon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4Cl2O2
CAS Number:
Molecular Weight:
227.04
Beilstein:
1073511
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

194-197 °C (lit.)

SMILES string

ClC1=C(Cl)C(=O)c2ccccc2C1=O

InChI

1S/C10H4Cl2O2/c11-7-8(12)10(14)6-4-2-1-3-5(6)9(7)13/h1-4H

InChI key

SVPKNMBRVBMTLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ahmad et al.
Archives of environmental contamination and toxicology, 29(4), 442-448 (1995-11-01)
Southern armyworm, Spodoptera eridania, larvae were provided ad libitum 0.002-0.25% w/w dichlone, 2,3-dichloro-1,4-naphthoquinone (CNQ). Larval mortality occurred in a time-and-dose dependent manner, with an LC17 of 0.01% and an LC50 of 0.26% CNQ at day-5. Extracts of larvae fed control
Inhibition of liver glycolysis in rats by dietary dichlone (2,3-dichloro-1,4-naphthoquinone).
C A Pritsos et al.
Bulletin of environmental contamination and toxicology, 35(1), 23-28 (1985-07-01)
Angupillai Satheshkumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 98, 378-383 (2012-09-18)
Various spectroscopy techniques (UV-Vis, DRS, FT-IR, (1)H NMR, LC-MS) and theoretical computations have been employed to investigate the mechanism of the nucleophilic substitution reaction of 2,3-dichloronaphthoquinone (DCNQ) with para-substituted anilines in solid state under base- and solvent-free conditions against traditional
Vishnu K Tandon et al.
European journal of medicinal chemistry, 44(3), 1086-1092 (2008-08-19)
A series of 2-chloro-3-arylsulfanyl-[1,4]naphthoquinones (2), 2,3-bis-arylsulfanyl-[1,4]naphthoquinones (3) and 12H-benzo[b]phenothiazine-6,11-diones and their analogs 6-8 were synthesized and evaluated for their antiproliferative activity against human cervical cancer (HeLa) cells. Compounds 3a and 3b were found to possess most potent antiproliferative and cell
Ibrahim A Darwish
Journal of AOAC International, 88(1), 38-45 (2005-03-12)
Three simple and sensitive spectrophotometric methods were developed and validated for determination of the hydrochloride salts of fluoxetine, sertraline, and paroxetine in their pharmaceutical dosage forms. These methods were based on the reaction of the N-alkylvinylamine formed from the interaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service