Skip to Content
Merck
All Photos(1)

Key Documents

D59100

Sigma-Aldrich

1,4-Dichlorobutane

99%

Synonym(s):

Tetramethylene dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$74.90
1 KG
$168.00

$74.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$74.90
1 KG
$168.00

About This Item

Linear Formula:
Cl(CH2)4Cl
CAS Number:
Molecular Weight:
127.01
Beilstein:
635673
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

161-163 °C (lit.)

mp

−38 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

ClCCCCCl

InChI

1S/C4H8Cl2/c5-3-1-2-4-6/h1-4H2

InChI key

KJDRSWPQXHESDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Dichlorobutane is extensively used in the synthesis of a variety of ligands, including commonly used 1,1′-(1,4-butanediyl)bis(imidazole) (bbi), to prepare coordination polymers and metal-organic frameworks (MOFs).[1][2][3] It can also be used a reagent in the synthesis of natural products such as spiro[4.5]-δ-damascone.[4]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A porous supramolecular architecture from a copper (II) coordination polymer with a 3D four-connected 86 net.
Ma JF, et al.
Inorganic Chemistry, 42(23), 7531-7534 (2003)
Synthesis and Olfactory Properties of a 6??Silasubstituted ?Spiro [4.5]?δ?Damascone?.
Lovchik M A and Kraft P
Chemistry?A European Journal , 23(19), 4590-4596 (2017)
Structures of metal-organic networks based on flexible 1, 1?-(1, 4-butanediyl) bis (imidazole-2-phenyl) ligand.
Liu YY, et al.
CrystEngComm, 10(5), 565-572 (2008)
Four new coordination polymers of Cu (II) with 1, 1?-(1, 4-butanediyl) bis (imidazole).
Ma JF, et al.
Polyhedron, 23(4), 553-559 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service