Skip to Content
Merck
All Photos(1)

Key Documents

D207705

Sigma-Aldrich

Diphenylfulvene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$222.00

$222.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$222.00

About This Item

Empirical Formula (Hill Notation):
C18H14
CAS Number:
Molecular Weight:
230.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$222.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

98%

form

solid

mp

81.5-83 °C (lit.)

SMILES string

c1ccc(cc1)C(\c2ccccc2)=C3\C=CC=C3

InChI

1S/C18H14/c1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16/h1-14H

InChI key

BULLHRADHZGONG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service