Skip to Content
Merck
All Photos(1)

Key Documents

D183202

Sigma-Aldrich

4,6-Dimethylpyrimidine

≥93%, technical grade

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$154.00

$154.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$154.00

About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
107068
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$154.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

grade

technical grade

Assay

≥93%

form

liquid

refractive index

n20/D 1.490 (lit.)

bp

154 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)ncn1

InChI

1S/C6H8N2/c1-5-3-6(2)8-4-7-5/h3-4H,1-2H3

InChI key

LSBIUXKNVUBKRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service