Skip to Content
Merck
All Photos(1)

Key Documents

D179604

Sigma-Aldrich

trans-2,5-Dimethylpiperazine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$110.00

$110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$110.00

About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

crystals

bp

162-165 °C (lit.)

mp

115-118 °C (lit.)

SMILES string

C[C@H]1CN[C@H](C)CN1

InChI

1S/C6H14N2/c1-5-3-8-6(2)4-7-5/h5-8H,3-4H2,1-2H3/t5-,6+

InChI key

NSMWYRLQHIXVAP-OLQVQODUSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Dermal - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romain Gautier et al.
Advanced materials (Deerfield Beach, Fla.), 31(14), e1807383-e1807383 (2019-02-19)
Hybrid metal halides containing perovskite layers have recently shown great potential for applications in solar cells and light-emitting diodes. Such compounds exhibit quantum confinement effects leading to tunable optical and electronic properties. Thus, broadband white-light emission has been observed from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service