D16207
4,4′-Diaminodiphenylamine sulfate salt
technical grade, 85%
Synonym(s):
4,4′-Iminodianiline sulfate salt
About This Item
Recommended Products
grade
technical grade
Assay
85%
mp
300 °C (lit.)
SMILES string
OS(O)(=O)=O.Nc1ccc(Nc2ccc(N)cc2)cc1
InChI
1S/C12H13N3.H2O4S/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12;1-5(2,3)4/h1-8,15H,13-14H2;(H2,1,2,3,4)
InChI key
OOZQLPDAELLDNY-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service