Skip to Content
Merck
All Photos(2)

Key Documents

D160008

Sigma-Aldrich

3,3-Dimethylglutarimide

99%

Synonym(s):

4,4-Dimethyl-2,6-piperidinedione

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$154.00

About This Item

Empirical Formula (Hill Notation):
C7H11NO2
CAS Number:
Molecular Weight:
141.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

mp

144-146 °C (lit.)

SMILES string

CC1(C)CC(=O)NC(=O)C1

InChI

1S/C7H11NO2/c1-7(2)3-5(9)8-6(10)4-7/h3-4H2,1-2H3,(H,8,9,10)

InChI key

YUJCWMGBRDBPDL-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Spiro-piperidine inhibitors for modulation of the M2 proton channel from influenza A virus
  • Second-generation selective inhibitors of Hepatitis C virus NS3 serine protease
  • Spirocyclic oxetanes
  • Ru-catalyzed stereoselective addition of imides to alkynes
  • Metabolically protected arylpiperiazine 5-HT1A ligands
  • Selective antagonists of the a1d adrenergic receptor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service