Skip to Content
Merck
All Photos(1)

Key Documents

D151009

Sigma-Aldrich

3,3′-Dimethylbiphenyl

99%

Synonym(s):

m,m′-Bitoluene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$176.00

$176.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$176.00

About This Item

Linear Formula:
CH3C6H4C6H4CH3
CAS Number:
Molecular Weight:
182.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$176.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.594 (lit.)

bp

286 °C/713 mmHg (lit.)

mp

5-7 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1)-c2cccc(C)c2

InChI

1S/C14H14/c1-11-5-3-7-13(9-11)14-8-4-6-12(2)10-14/h3-10H,1-2H3

InChI key

GVEDOIATHPCYGS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service