Skip to Content
Merck
All Photos(1)

Key Documents

D120405

Sigma-Aldrich

4,6-Dihydroxypyrimidine

98%

Synonym(s):

4,6-Pyrimidinediol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$28.30

$28.30

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$28.30

About This Item

Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
Beilstein:
112224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$28.30

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

98%

form

powder

mp

>300 °C (lit.)

SMILES string

Oc1cc(O)ncn1

InChI

1S/C4H4N2O2/c7-3-1-4(8)6-2-5-3/h1-2H,(H2,5,6,7,8)

InChI key

DUFGYCAXVIUXIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is it the alpha or the beta form of the compound that is available. Is there any Powder XRD data available for the compound?

    1 answer
    1. There is no XRD data or other crystallographic information available for this item.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service