Key Documents
CLS998565
Pyrex® watch glasses
O.D. 65 mm
Synonym(s):
Corning® 9985
Select a Size
144 EA
$420.00
$420.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
144 EA
$420.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$420.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
Corning 9985-65
O.D.
65 mm
General description
Heavy-wall, borosilicate glass with fire-polished edge provides durability. May be slightly out-of-round due to O.D. variability.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)