Key Documents
CLS70792N
Pyrex® serological pipettes, disposable, non-sterile, unplugged
volume 2.0 mL
Synonym(s):
pipet
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
Pricing and availability is not currently available.
material
borosilicate glass
sterility
non-sterile
feature
DNase free: no
RNase free: no
graduations
subdivision
packaging
case of 700 ea
manufacturer/tradename
Corning 7079-2N
volume measuring range
2.0 mL
capacity
2.0 mL
tip size
, Narrow
Related Categories
Features and Benefits
  • Disposable PYREX® glass pipettes are calibrated "to deliver" (TD) with blow-out
  • Long, slender, tapered tips of pipettes make pipetting faster and easier
  • All pipettes have negative graduations and fire polished tips for burr-free, even flow rates
  • Nonsterile, unplugged and bulk packed.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)